4-Dodecylanilin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Dodecylphenylamine, 1-Amino-4-dodecylbenzene
M-Clarity™ Qualitätsstufe = MQ100
233544-5G
SIAL233544-5GEA
147
EUR
SIAL233544-5G
4-Dodecylanilin, Sigma-Aldrich®
4-Dodecylanilin
Formel:
C₁₈H₃₁N Molecular Weight: 261,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007919 CAS-Nummer: 104-42-7 EINECS: 203-201-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...