4-Amino-2-mercaptopyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine

M-Clarity™ Qualitätsstufe = MQ100

233625-1G
SIAL233625-1GEA 58.5 EUR
SIAL233625-1G
4-Amino-2-mercaptopyrimidin, Sigma-Aldrich®
4-Amino-2-mercaptopyrimidine
Formel: C₄H₅N₃S
Molecular Weight: 127,1676 g/mol
MDL: MFCD00006075
CAS-Nummer: 333-49-3
EINECS: 206-374-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR