2-Chlorphenylphosphorodichloridat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

235237-25G
SIAL235237-25GEA 83.7 EUR
SIAL235237-25G
2-Chlorphenylphosphorodichloridat, Sigma-Aldrich®
2-Chlorphenylphosphorodichloridat
Formel: C₆H₄Cl₃O₂P
Molecular Weight: 245,43 g/mol
MDL: MFCD00009704
CAS-Nummer: 15074-54-1
EINECS: 239-129-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR