3,6-Dithia-1,8-octandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,8-Dihydroxy-3,6-Dithiaoctane, 2,2-(Ethane-1,2-diylbis(sulphanediyl))diethanol

M-Clarity™ Qualitätsstufe = MQ200

235334-25G
SIAL235334-25GEA 66.8 EUR
SIAL235334-25G
3,6-Dithia-1,8-octandiol, Sigma-Aldrich®
3,6-Dithia-1,8-octandiol
Formel: C₆H₁₄O₂S₂
Molecular Weight: 182,31 g/mol
MDL: MFCD00002911
CAS-Nummer: 5244-34-8
EINECS: 226-045-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR