2,4-Dichlor-5-nitrobenzotrifluorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2,4-Dichloro-5-nitrobenzotrifluoride, 1,3-Dichloro-5-nitro-4-(trifluoromethyl)benzene
M-Clarity™ Qualitätsstufe = MQ100
235814-25G
SIAL235814-25GEA
47
EUR
SIAL235814-25G
2,4-Dichlor-5-nitrobenzotrifluorid, Sigma-Aldrich®
2,4-Dichlor-5-nitrobenzotrifluorid
Formel:
C₇H₂Cl₂F₃NO₂ Molecular Weight: 260 g/mol Schmelzpunkt: 59,5 °C |
MDL:
MFCD00007073 CAS-Nummer: 400-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...