Brij® L4 average Mn ~362, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Brij 30, Laureth-4, Brij L4

MQ200

235989-100ML 235989-1L 235989-5ML
SIAL235989-100MLEA 37.1 EUR
SIAL235989-100ML SIAL235989-1L SIAL235989-5ML
Brij® L4 average Mn ~362, Sigma-Aldrich®
Brij® L4
Formel: (C₂₀H₄₂O₅)n
MDL: MFCD00043063
CAS-Nummer: 9002-92-0
EINECS: 500-002-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR