1,2,4,5-Tetrakis(brommethyl)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

236616-5G
SIAL236616-5GEA 201 EUR
SIAL236616-5G
1,2,4,5-Tetrakis(brommethyl)benzol, Sigma-Aldrich®
1,2,4,5-Tetrakis(brommethyl)benzol
Formel: C₁₀H₁₀Br₄
Molecular Weight: 449,8 g/mol
MDL: MFCD00009664
CAS-Nummer: 15442-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR