Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')lanthan(III), Sigma-Aldrich®

Lieferant: Merck

Synonyme: La(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)

237264-1G
SIAL237264-1GEA 82.6 EUR
SIAL237264-1G
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')lanthan(III), Sigma-Aldrich®
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')lanthan(III)
Formel: C₃₃H₅₇LaO₆
Molecular Weight: 688,71 g/mol
MDL: MFCD00010466
CAS-Nummer: 14319-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR