Dimethyl-5-nitroisophthalat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Nitro-1,3-benzoldicarbonsäuredimethylester, 5-Nitroisophthalsäuredimethylester

M-Clarity™ Qualitätsstufe = MQ100

237361-25G
SIAL237361-25GEA 51 EUR
SIAL237361-25G
Dimethyl-5-nitroisophthalat, Sigma-Aldrich®
Dimethyl-5-nitroisophthalat
Formel: C₁₀H₉NO₆
Molecular Weight: 239,18 g/mol
MDL: MFCD00008429
CAS-Nummer: 13290-96-5
EINECS: 236-307-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR