(-)-DIOP, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R,R)-DIOP, (4R,5R)-(-)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (((4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)

M-Clarity™ Qualitätsstufe = MQ100

237655-1G
SIAL237655-1GEA 340 EUR
SIAL237655-1G
(-)-DIOP, Sigma-Aldrich®
(-)-DIOP
Formel: C₃₁H₃₂O₂P₂
Molecular Weight: 498,532 g/mol
MDL: MFCD00014107
CAS-Nummer: 32305-98-9
EINECS: 250-984-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR