Ammoniumtrifluoracetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trifluoressigsäure Ammoniumsalz

M-Clarity™ Qualitätsstufe = MQ200

238732-25G 238732-100G
SIAL238732-25GEA 72.7 EUR
SIAL238732-25G SIAL238732-100G
Ammoniumtrifluoracetat, Sigma-Aldrich®
Ammoniumtrifluoracetat
Formel: C₂H₄F₃NO₂
Molecular Weight: 131.05 g/mol
MDL: MFCD00012615
CAS-Nummer: 3336-58-1
EINECS: 222-072-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR