N,N'-Bis(3-aminopropyl)ethylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,5,8,12-Tetraazadodecane

M-Clarity™ Qualitätsstufe = MQ100

239399-100ML
SIAL239399-100MLEA 34.5 EUR
SIAL239399-100ML
N,N'-Bis(3-aminopropyl)ethylendiamin, Sigma-Aldrich®
N,N'-Bis(3-aminopropyl)ethylendiamin
Formel: C₈H₂₂N₄
Molecular Weight: 174.29 g/mol
MDL: MFCD00008210
CAS-Nummer: 10563-26-5
EINECS: 234-147-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR