1,4-Bis(3-aminopropyl)piperazine ≥99%, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,4-Di(3-aminopropyl)piperazine
M-Clarity™ Qualitätsstufe = MQ100
239488-50ML
SIAL239488-50MLEA
122
EUR
SIAL239488-50ML
1,4-Bis(3-aminopropyl)piperazine ≥99%, Sigma-Aldrich®
1,4-Bis(3-aminopropyl)piperazine
1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K).
The thermodynamic parameters for protonation of 1,4-bis(3-aminopropyl)-piperazine (BAPP) were studied in aqueous solution using a potentiometric technique.
Formel:
C₁₀H₂₄N₄ Molecular Weight: 200,33 g/mol Dichte: 0,973 g/ml (25 °C) (Lit.) |
MDL:
MFCD00006161 CAS-Nummer: 7209-38-3 |
Spezifikation Testergebnisse
CAS Number | 7209-38-3 |
Empirical Formula (Hill Notation) | C₁₀H₂₄N₄ |
Molecular Weight | 200,32 |
EC Number | 230-589-1 |
MDL number | MFCD00006161 |
PubChem Substance ID | 24854378 |
Vapor density | 6,9 (vs air) |
InChI Key | XUSNPFGLKGCWGN-UHFFFAOYSA-N |
Assay | ≥99% |
Refractive index | n20/D 1,502(lit.) |
Bp | 150 - 152 °C/2 mm Hg(lit.) |
Mp | 15 °C(lit.) |
Density | 0,973 g/ml at 25 °C(lit.) |
Gene Information | rat ... Grin2a(24409) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...