trans-Benzylidenaceton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-4-Phenylbut-3-en-2-one, trans-4-Phenyl-3-buten-2-one

M-Clarity™ Qualitätsstufe = MQ100

241091-50G
SIAL241091-50GEA 62 EUR
SIAL241091-50G
trans-Benzylidenaceton, Sigma-Aldrich®
trans-Benzylidenaceton
Formel: C₁₀H₁₀O
Molecular Weight: 146,19 g/mol
MDL: MFCD00008779
CAS-Nummer: 1896-62-4
EINECS: 217-587-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR