2,2'-Bithiophen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2′-Dithienyl, 2,2′-Bithienyl

241636-10G
SIAL241636-10GEA 202 EUR
SIAL241636-10G
2,2'-Bithiophen, Sigma-Aldrich®
2,2'-Bithiophen
Formel: C₈H₆S₂
Molecular Weight: 166,27 g/mol
MDL: MFCD00005414
CAS-Nummer: 492-97-7
EINECS: 207-767-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR