(R)-(+)-N,N-Dimethyl-1-phenylethanamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-N,N-Dimethyl-1-phenylethanamine

242071-1G
SIAL242071-1GEA 87.1 EUR
SIAL242071-1G
(R)-(+)-N,N-Dimethyl-1-phenylethanamin, Sigma-Aldrich®
(R)-(+)-N,N-Dimethyl-1-phenylethanamin
Formel: C₁₀H₁₅N
Molecular Weight: 149,230 g/mol
MDL: MFCD00008857
CAS-Nummer: 19342-01-9
EINECS: 242-976-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR