2-Chlor-p-phenylendiaminsulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlorobenzene-1,4-diamine sulphate, 2-Chloro-1,4-phenylenediammonium sulphate

24240-100G
SIAL24240-100GEA 38.6 EUR
SIAL24240-100G
2-Chlor-p-phenylendiaminsulfat, Sigma-Aldrich®
2-Chlor-p-phenylendiaminsulfat
Formel: C₆H₉ClN₂O₄S
Molecular Weight: 240.67 g/mol
MDL: MFCD00013004
CAS-Nummer: 61702-44-1
EINECS: 262-915-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR