(Brommethyl)cyclopropan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

242403-1G 242403-5G
SIAL242403-1GEA 51 EUR
SIAL242403-1G SIAL242403-5G
(Brommethyl)cyclopropan, Sigma-Aldrich®
(Brommethyl)cyclopropan
Formel: C₄H₇Br
Molecular Weight: 135 g/mol
Siedepunkt: 105
Dichte: 1.41 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00001306
CAS-Nummer: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR