4-Acetoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Formylphenylacetat

M-Clarity™ Qualitätsstufe = MQ100

242608-10G
SIAL242608-10GEA 95 EUR
SIAL242608-10G
4-Acetoxybenzaldehyd, Sigma-Aldrich®
4-Acetoxybenzaldehyd
Formel: C₉H₈O₃
Molecular Weight: 164,16 g/mol
MDL: MFCD00003384
CAS-Nummer: 878-00-2
EINECS: 212-898-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR