2-Chlorethyl-methylsulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Chloroethyl)(methyl)sulphane

M-Clarity™ Qualitätsstufe = MQ200

242632-5G
SIAL242632-5GEA 79.2 EUR
SIAL242632-5G
2-Chlorethyl-methylsulfid, Sigma-Aldrich®
2-Chlorethyl-methylsulfid
Formel: C₃H₇ClS
Molecular Weight: 110.61 g/mol
MDL: MFCD00000978
CAS-Nummer: 542-81-4
EINECS: 208-828-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR