Ethylanthranilat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminobenzoesäureethylester, Ethyl-2-aminobenzoat, Anthranilsäureethylester

242977-100G
SIAL242977-100GEA 50.6 EUR
SIAL242977-100G
Ethylanthranilat, Sigma-Aldrich®
Ethylanthranilat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Siedepunkt: 268 °C (1013 hPa)
Dichte: 1,12 g/cm³ (20 °C)
MDL: MFCD00007711
CAS-Nummer: 87-25-2
EINECS: 201-735-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR