2-sec-Butyl-3-methoxypyrazin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(sec-Butyl)-3-methoxypyrazine
M-Clarity™ Qualitätsstufe = MQ100
243116-1G
SIAL243116-1GEA
34.5
EUR
SIAL243116-1G
2-sec-Butyl-3-methoxypyrazin, Sigma-Aldrich®
2-sec-Butyl-3-methoxypyrazin
Formel:
C₉H₁₄N₂O Molecular Weight: 166,2203 g/mol |
MDL:
MFCD00006136 CAS-Nummer: 24168-70-5 EINECS: 246-050-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...