(R)-(-)-Methyl-3-hydroxybutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate

M-Clarity™ Qualitätsstufe = MQ100

243159-1G
SIAL243159-1GEA 95.3 EUR
SIAL243159-1G
(R)-(-)-Methyl-3-hydroxybutyrat, Sigma-Aldrich®
(R)-(-)-Methyl-3-hydroxybutyrat
Formel: C₅H₁₀O₃
Molecular Weight: 118,13 g/mol
MDL: MFCD00063289
CAS-Nummer: 3976-69-0
EINECS: 223-610-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR