(S)-(+)-Methyl-3-hydroxybutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl (S)-(+)-3-Hydroxybutyrate, (S)-Methyl 3-hydroxybutanoate, Methyl (S)-(+)-3-hydroxybutyrate

M-Clarity™ Qualitätsstufe = MQ100

243167-1G
SIAL243167-1GEA 45.5 EUR
SIAL243167-1G
(S)-(+)-Methyl-3-hydroxybutyrat, Sigma-Aldrich®
(S)-(+)-Methyl-3-hydroxybutyrat
Formel: C₅H₁₀O₃
Molecular Weight: 118,13 g/mol
MDL: MFCD00064461
CAS-Nummer: 53562-86-0
EINECS: 258-628-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR