2-Chlor-N,N-dimethylacetamid, Sigma-Aldrich®

Lieferant: Merck

24350-25ML-F
SIAL24350-25ML-FEA 248 EUR
SIAL24350-25ML-F
2-Chlor-N,N-dimethylacetamid, Sigma-Aldrich®
2-Chlor-N,N-dimethylacetamid
Formel: C₄H₈ClNO
Molecular Weight: 121.57 g/mol
MDL: MFCD00039360
CAS-Nummer: 2675-89-0
EINECS: 220-224-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR