Molybdändisilicid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Molybdenum silicide
M-Clarity™ Qualitätsstufe = MQ100
243647-250G
SIAL243647-250GEA
300
EUR
SIAL243647-250G
Molybdändisilicid, Sigma-Aldrich®
Molybdenum disilicide
Formel:
MoSi₂ Molecular Weight: 152.11 g/mol |
MDL:
MFCD00151535 CAS-Nummer: 12136-78-6 EINECS: 235-231-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...