cis-Bis-(triphenylphosphin)-platin(II)-dichlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
cis-Bis(triphenylphosphin)palladium(II)chlorid
M-Clarity™ Qualitätsstufe = MQ100
244945-1G
SIAL244945-1GEA
156
EUR
SIAL244945-1G
cis-Bis-(triphenylphosphin)-platin(II)-dichlorid, Sigma-Aldrich®
cis-Bis-(triphenylphosphin)-platin(II)-dichlorid
Formel:
[(C₆H₅)₃P]₂PtCl₂ Molecular Weight: 790,56 g/mol |
MDL:
MFCD00010825 CAS-Nummer: 15604-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...