2,2,6-Trimethyl-4H-1,3-dioxin-4-on, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,6-Trimethyl-1,3-dioxine-4-one

M-Clarity™ Qualitätsstufe = MQ200

245100-100G
SIAL245100-100GEA 96.2 EUR
SIAL245100-100G
2,2,6-Trimethyl-4H-1,3-dioxin-4-on, Sigma-Aldrich®
2,2,6-Trimethyl-4H-1,3-dioxin-4-on
Formel: C₇H₁₀O₃
Molecular Weight: 142,15 g/mol
MDL: MFCD00040468
CAS-Nummer: 5394-63-8
EINECS: 226-403-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR