Methyl-4-chloracetoacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-chloro-3-oxobutanoate

M-Clarity™ Qualitätsstufe = MQ100

245860-5G
SIAL245860-5GEA 78.2 EUR
SIAL245860-5G
Methyl-4-chloracetoacetat, Sigma-Aldrich®
Methyl-4-chloracetoacetat
Formel: C₅H₇ClO₃
Molecular Weight: 150,56 g/mol
MDL: MFCD00000938
CAS-Nummer: 32807-28-6
EINECS: 251-230-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR