4-Phenoxybenzoesäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
246182-25G
SIAL246182-25GEA
80.1
EUR
SIAL246182-25G
4-Phenoxybenzoesäure, Sigma-Aldrich®
4-Phenoxybenzoesäure
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214,22 g/mol |
MDL:
MFCD00002539 CAS-Nummer: 2215-77-2 EINECS: 218-682-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...