6-Chlorindol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Chloro-1H-indole
M-Clarity™ Qualitätsstufe = MQ100
246239-1G
SIAL246239-1GEA
64.8
EUR
SIAL246239-1G
6-Chlorindol, Sigma-Aldrich®
6-Chlorindol
Formel:
C₈H₆ClN Molecular Weight: 151,6 g/mol Schmelzpunkt: 90 °C |
MDL:
MFCD00005681 CAS-Nummer: 17422-33-2 EINECS: 241-449-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...