4-Hydrazinobenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

246395-25G 246395-5G
SIAL246395-25GEA 72.7 EUR
SIAL246395-25G SIAL246395-5G
4-Hydrazinobenzoesäure, Sigma-Aldrich®
4-Hydrazinobenzoic acid
Formel: C₇H₈N₂O₂
MDL: MFCD00007581
CAS-Nummer: 619-67-0
EINECS: 210-609-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR