trans,trans-Dibenzylidenaceton, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

246425-50G
SIAL246425-50GEA 196 EUR
SIAL246425-50G
trans,trans-Dibenzylidenaceton, Sigma-Aldrich®
trans,trans-Dibenzylidenaceton
Formel: C₁₇H₁₄O
MDL: MFCD00004790
CAS-Nummer: 35225-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR