Chlordimethyloctylsilan, Sigma-Aldrich®

Lieferant: Merck

246859-25G
SIAL246859-25GEA 134 EUR
SIAL246859-25G
Chlordimethyloctylsilan, Sigma-Aldrich®
Chlordimethyloctylsilan
Formel: C₁₀H₂₃ClSi
MDL: MFCD00000511
CAS-Nummer: 18162-84-0
EINECS: 242-044-5
UN: 2987
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR