1,8-Bis(brommethyl)naphthalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

246921-250MG
SIAL246921-250MGEA 187 EUR
SIAL246921-250MG
1,8-Bis(brommethyl)naphthalin, Sigma-Aldrich®
1,8-Bis(brommethyl)naphthalin
Formel: C₁₂H₁₀Br₂
MDL: MFCD00010251
CAS-Nummer: 2025-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR