4-Amino-3-nitrobenzoesäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Amino-3-nitrobenzoic Acid
M-Clarity™ Qualitätsstufe = MQ100
248118-25G
SIAL248118-25GEA
39.1
EUR
SIAL248118-25G
4-Amino-3-nitrobenzoesäure, Sigma-Aldrich®
4-Amino-3-nitrobenzoic acid
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol Schmelzpunkt: 290 °C |
MDL:
MFCD00017009 CAS-Nummer: 1588-83-6 EINECS: 216-453-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...