3-Methyl-2-thenoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methylthiophene-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

248207-5G
SIAL248207-5GEA 22.3 EUR
SIAL248207-5G
3-Methyl-2-thenoesäure, Sigma-Aldrich®
3-Methyl-2-thenoesäure
Formel: C₆H₆O₂S
Molecular Weight: 142,18 g/mol
MDL: MFCD00005438
CAS-Nummer: 23806-24-8
EINECS: 245-894-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR