5-Norbornene-2-methanol (endo + exo Gemisch) ≥98%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Bicyclo[2.2.1]hept-5-ene-2-methanol, 5-Norbornen-2-methanol (mixture of isomers), 2-Hydroxymethyl-5-norbornene, Bicyclo[2.2.1]hept-5-en-2-ylmethanol

M-Clarity™ Qualitätsstufe = MQ100

248533-5G
SIAL248533-5GEA 115 EUR
SIAL248533-5G
5-Norbornene-2-methanol (endo + exo Gemisch) ≥98%, Sigma-Aldrich®
5-Norbornene-2-methanol

5-Norbornene-2-methanol has been used in the preparation of 4-(norborn-2-ene-5-ylmethyleneoxy)-2,2′:6′,2′′-terpyridine and 5- {[(3,5-di-t-butyl-4-hydroxy-benzoyl)oxy]methyl}-2-norbornene (BHBN).


Co-polymerization of 5-norbornene-2-methanol with ethylene using heterogeneous silica supported catalysts has been reported.

Formel: C₈H₁₂O
Molecular Weight: 124,183 g/mol
Siedepunkt: 97 °C (20 mmHg) (l)
Dichte: 1,027 g/ml (25 °C)
Flammpunkt: 87 °C
MDL: MFCD00167571
CAS-Nummer: 95-12-5

Bestellen

Spezifikation Testergebnisse

Assay 98%
Refractive index n20/D 1,500 (l)
Density (25 °C) 1,027 g/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR