(Brommethyl)trimethylsilan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
249777-5G
SIAL249777-5GEA
73
EUR
SIAL249777-5G
(Brommethyl)trimethylsilan, Sigma-Aldrich®
(Bromomethyl)trimethylsilane
Formel:
BrH₂CSi(CH₃)₃ Siedepunkt: 116 °C (1013 hPa) Dichte: 1,17 g/cm³ (25 °C) |
MDL:
MFCD00000167 CAS-Nummer: 18243-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...