(±)-2-Bromisovaleriansäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-3-methylbutyricacid, 2-Bromo-3-methylbutyric acid

M-Clarity™ Qualitätsstufe = MQ200

251003-25G
SIAL251003-25GEA 108 EUR
SIAL251003-25G
(±)-2-Bromisovaleriansäure, Sigma-Aldrich®
(±)-2-Bromisovaleriansäure
Formel: C₅H₉BrO₂
Molecular Weight: 181,03 g/mol
MDL: MFCD00004210
CAS-Nummer: 565-74-2
EINECS: 209-291-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR