2-Hydroxynicotinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

251054-100G 251054-25G
SIAL251054-100GEA 109 EUR
SIAL251054-100G SIAL251054-25G
2-Hydroxynicotinsäure, Sigma-Aldrich®
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR