2-(Methylthio)nicotinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

251089-25G
SIAL251089-25GEA 160 EUR
SIAL251089-25G
2-(Methylthio)nicotinsäure, Sigma-Aldrich®
2-(Methylthio)nicotinsäure
Formel: C₇H₇NO₂S
Molecular Weight: 169,20 g/mol
MDL: MFCD00010101
CAS-Nummer: 74470-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR