(S)-(+)-Methylmandelat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(+)-2-Hydroxy-2-phenylethansäuremethylester, L(+)-Methylglykolat, L(+)-2-Hydroxy-2-phenylessigsäuremethylester, Methyl-L(+)-glykolat, (S)-(+)-Methylglykolat, (S)-(+)-2-Hydroxy-2-phenylessigsäuremethylester, L-2-Hydroxy-2-phenylethansäuremethylester, L(+)-Mandelsäuremethylester, L(+)-Methylmandelat, Methyl-(S)-(+)-glykolat, (S)-(+)-Mandelsäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

251542-5G
SIAL251542-5GEA 27.1 EUR
SIAL251542-5G
(S)-(+)-Methylmandelat, Sigma-Aldrich®
(S)-(+)-Methylmandelat
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Siedepunkt: 100…105 °C (1,25 mmHg)
Schmelzpunkt: 58 °C
MDL: MFCD00064246
CAS-Nummer: 21210-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR