Kalium-3-(methacryloyloxy)-1-propansulfonat 96,0-104,0 Titrimetric, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Methacrylic acid 3-sulphopropyl Ester Potassium salt, 3-sulphopropyl Methacrylate Potassium Salt, Potassium 3-(Methacryloyloxy)propanesulphonate

MQ100

251631-100G
SIAL251631-100GEA 91 EUR
SIAL251631-100G
Kalium-3-(methacryloyloxy)-1-propansulfonat 96,0-104,0 Titrimetric, Sigma-Aldrich®
Kalium-3-(methacryloyloxy)-1-propansulfonat

The monomer was polymerized to prepare a hydrogel test bed, which was used in a study an in vitro wound infection model. It was polymerized with N-isopropyl acrylamide to prepare thermoresponsive super water absorbent hydrogels, superporous hydrogels (SPH) and SPH based composites.

Formel: C₇H₁₂O₅S
Molecular Weight: 246.32 g/mol
Schmelzpunkt: 302 °C (Lit.)
MDL: MFCD00010139
CAS-Nummer: 31098-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR