Zinc meso-tetraphenylporphine ≥94,0% chlorarm, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,10,15,20-Tetraphenyl-21H,23H-porphine Zinc(II), Zinc(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine zinc, Zinc TPP, Zinc(II) TPP, Tetraphenylporphine zinc, TPP Zn

M-Clarity™ Qualitätsstufe = MQ100

252174-25MG
SIAL252174-25MGEA 0 EUR
SIAL252174-25MG
Zinc meso-tetraphenylporphine ≥94,0% chlorarm, Sigma-Aldrich®
Zinc meso-tetraphenylporphine
Formel: C₄₄H₂₈N₄Zn
Molecular Weight: 678,10 g/mol
Dichte: 1,276 g/cm³ (20 °C)
MDL: MFCD00012155
CAS-Nummer: 14074-80-7
EINECS: 213-025-9

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Purple to Violet
Appearance (Form) Powder, Crystals or Crystalline Powder
Infrared Spectrum Conforms to Structure
Wavelength Approximately 421 nm
Extinction Coefficient ≥ 400000 (c = 0.002g/l, Ether)
Carbon 73.3 - 82.6 %
Nitrogen 7.7 - 8.8 %
Purity (HPLC) ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR