2-Brombiphenyl (PBB Nr. 1), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Brom-2-phenylbenzol, PBB Nr. 1
M-Clarity™ Qualitätsstufe = MQ100
252247-5G
SIAL252247-5GEA
102
EUR
SIAL252247-5G
2-Brombiphenyl (PBB Nr. 1), Sigma-Aldrich®
2-Brombiphenyl (PBB Nr. 1)
Formel:
C₁₂H₉Br Molecular Weight: 233,11 g/mol Siedepunkt: 297 °C (1013 hPa) Schmelzpunkt: 2 °C Dichte: 1,2766 g/cm³ (20 °C) |
MDL:
MFCD00000065 CAS-Nummer: 2052-07-5 EINECS: 218-141-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...