4-Hydroxybutanon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-2-butanone, 4-Hydroxybutan-2-one

M-Clarity™ Qualitätsstufe = MQ100

252557-100G
SIAL252557-100GEA 160 EUR
SIAL252557-100G
4-Hydroxybutanon, Sigma-Aldrich®
4-Hydroxybutanon
Formel: C₄H₈O₂
Molecular Weight: 88,11 g/mol
MDL: MFCD00059005
CAS-Nummer: 590-90-9
EINECS: 209-693-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR