Trimethylsilylisocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

252646-25G 252646-5G
SIAL252646-25GEA 243 EUR
SIAL252646-25G SIAL252646-5G
Trimethylsilylisocyanat, Sigma-Aldrich®
Trimethylsilylisocyanat
Formel: C₄H₉NOSi
MDL: MFCD00001993
CAS-Nummer: 1118-02-1
EINECS: 214-256-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR