(R)-(-)-2-Methyl-2,4-pentandiol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

252840-1G
SIAL252840-1GEA 126 EUR
SIAL252840-1G
(R)-(-)-2-Methyl-2,4-pentandiol, Sigma-Aldrich®
(R)-(-)-2-Methyl-2,4-pentandiol
Formel: C₆H₁₄O₂
Molecular Weight: 118,18 g/mol
MDL: MFCD00064275
CAS-Nummer: 99210-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR