Tris(diethylamino)phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hexaethylphosphortriamid

M-Clarity™ Qualitätsstufe = MQ200

253189-25G 253189-5G
SIAL253189-25GEA 193 EUR
SIAL253189-25G SIAL253189-5G
Tris(diethylamino)phosphin, Sigma-Aldrich®
Tris(diethylamino)phosphin
Formel: C₁₂H₃₀N₃P
Molecular Weight: 247.36 g/mol
Dichte: 0.89 g/cm³ (20 °C)
MDL: MFCD00009041
CAS-Nummer: 2283-11-6
EINECS: 218-920-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR